Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibodies Inc
For maximum recovery of product, centrifuge the vial prior to removing the cap.
SYT9 may be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ45896; synaptogamin V; Synaptotagmin 5; synaptotagmin 9; Synaptotagmin IX; Synaptotagmin V; Synaptotagmin-9; Synaptotagmin9; SynaptotagminIX; Syt IX; SYT-9; SytIX
Gene Aliases: Syt5; SYT9; Sytv
UniProt ID: (Human) Q86SS6, (Mouse) Q9R0N9, (Rat) Q925C0
Entrez Gene ID: (Human) 143425, (Mouse) 60510, (Rat) 60564
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support