Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Despite its predicted molecular weight, Syntaphilin often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human brain tissue lysate.
PA5-20528 can be used with blocking peptide PEP-0648.
Syntaphilin was initially identified in a yeast two-hybrid screen with the carboxy terminal region of Syntaxin-1 as bait. Syntaxin-1 is a key component of the synaptic vesicle docking machinery that forms the SNARE complex with synaptobrevin and SNAP-25. Syntaphilin competes with SNAP-25 for binding to syntaxin-1 and inhibits the formation of the SNARE complex, thereby potentially regulating synaptic vesicle exocytosis. Syntaphilin also binds dynamin-1 and inhibits dynamin-dependent endocytosis. Mice lacking syntaphilin show an increased level of mitochondrial motility and a reduced density of axonal mitochondria. This correlates with an enhanced short-term facilitation and significant impairments in motor ability, suggesting syntaphilin plays a major role in presynaptic function. Multiple isoforms are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bA314N13.5; Syntaphilin
Gene Aliases: 6430515A01; AW045671; AW556958; AW743098; KIAA0374; mKIAA0374; SNPH
UniProt ID: (Human) O15079, (Rat) B5DF41, (Mouse) Q80U23
Entrez Gene ID: (Human) 9751, (Rat) 296267, (Mouse) 241727
Molecular Function:
SNARE protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support