Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAPDPWFST YDSTCQIAQE IAEKIQQRNQ YERKGEKAPK LTVTIRALLQ NLKEKIALLK DLLLRAVSTH QITQLEGDRR QNLLDDLVTR ERLLLASFKN EGAEPDLIRS SLMSEEAKRG APNPWLFEEP EETRGLGFDE IRQQQQKIIQ EQDAGLDALS SIISRQKQMG QEIGNELDEQ NEIIDDLANL VENTDEKLRN ETRRVNMVDR KS (1-211 aa encoded by BC009713)
The gene is a member of the syntaxin family. The encoded protein is involved in protein trafficking from early to late endosomes via vesicle fusion and exocytosis. A related pseudogene has been identified on chromosome 12. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CIP-1-associated regulator of cyclin B; Syntaxin-8; Syntaxin-like protein 3I35
Gene Aliases: 0610007H08Rik; 1110002H11Rik; 4930571E13Rik; CARB; STX8
UniProt ID: (Human) Q9UNK0, (Rat) Q9Z2Q7, (Mouse) O88983
Entrez Gene ID: (Human) 9482, (Rat) 59074, (Mouse) 55943
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support