Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Antibody is stable for 24 months.
Positive Control: HeLa or 293T cells.Human testis, spleen or colon. Cellular Localization: Nucleus.
Specificity Comments: TAF42 (SPT3-associated factor 42), also known as TADA1 (transcriptional adapter 1), ADA1, HFI1 or hADA1, is a 335 amino acid nuclear protein that belongs to the TADA1 family. As a component of the STAGA transcription coactivator-HAT complex, STAF42 is most likely involved in transcriptional regulation. The gene that encodes STAF42 maps to human chromosome 1, which spans 260 million base pairs, contains over 3,000 genes and comprises nearly 8% of the human genome. Chromosome 1 houses a large number of disease associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson s disease, Gaucher disease, schizophrenia and Usher syndrome. Aberrations in chromosome 1 are found in a variety of cancers, including head and neck cancer, malignant melanoma and multiple myeloma.
Predicted to react with Rabbit, Goat, Horse, Guinea pig, Dog, Pig, Rhesus monkey, Orangutan and Elephant (based on sequence homology)
Probably involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HF1L; SPT3-associated factor 42; SPT3-associated factor 42 (STAF42); STAF42; Transcriptional adapter 1; Transcriptional adapter 1-like protein; transcriptional adaptor 1 (HFI1 homolog, yeast)
Gene Aliases: ADA1; hADA1; HFI1; STAF42; TADA1; TADA1L
UniProt ID: (Human) Q96BN2
Entrez Gene ID: (Human) 117143
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support