Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MSELKDCPLQ FHDFKSVDHL KVCPRYTAVL ARSEDDGIGI EELDTLQLEL ETLLSSASRR LRVLEAETQI LTDWQDKKGD RRFLKLGRDH ELGAPPKHGK PKKQKLEGKA GHGPGPGPGR PKSKNLQPKI QEYEFTDDPI DVPRIPKNDA PNRFWASVEP YCADITSEEV RTLEELLKPP EDEAEHYKIP PLGKHYSQRW AQEDLLEEQK DGARAAAVAD KKKGLMGPLT ELDTKDVDAL LKKSEAQHEQ PEDGCPFGAL TQRLLQALVE ENIISPMEDS PIPDMSGKES GADGASTSPR NQNKPFSVPH TKSLESRIKE ELIAQGLLES EDRPAEDSED EVLAELRKRQ AELKALSAHN RTKKHDLLRL AKEEVSRQEL RQRVRMADNE VMDAFRKIMA ARQKKRTPTK KEKDQAWKTL KERESILKLL DG
Many DNA-binding transcriptional activator proteins enhance the initiation rate of RNA polymerase II-mediated gene transcription by interacting functionally with the general transcription machinery bound at the basal promoter. Adaptor proteins are usually required for this activation, possibly to acetylate and destabilize nucleosomes, thereby relieving chromatin constraints at the promoter. The protein encoded by this gene is a transcriptional activator adaptor and has been found to be part of the PCAF histone acetylase complex. In addition, it associates with the tumor suppressor protein p53 and is required for full activity of p53 and p53-mediated apoptosis. At least four alternatively spliced variants have been found for this gene, but the full-length nature of some variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADA3 homolog; ADA3-like protein; alteration/deficiency in activation 3; FLJ20221; FLJ21329; hADA3; STAF54; Transcriptional adapter 3; Transcriptional adapter 3-like
Gene Aliases: ADA3; hADA3; NGG1; STAF54; TADA3; TADA3L
UniProt ID: (Human) O75528
Entrez Gene ID: (Human) 10474
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support