Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: LAVQNSSLHD IQHAPKLSAL KQLLLDCGLG NGSTSESGTE SVVAQHRILI FCQLKSMLDI VEHDLLKPHL PSVTYLRLDG SIPPGQRHSI VSRFNNDPSI DVLLLTTHVG GLGLNLTGAD TVVFVEHDWN PMRDLQAMDR AHRIGQKRVV NVYRLITRGT LEEKIMGLQK FKMNIANTVI SQENSSLQSM GTDQLLDLFT LDKDGKAEKA DTSTSGKASM KSILENLSDL WDQEQYDSEY SLENFMHSLK; Positive Samples: HepG2, HeLa, MCF7, Rat testis; Cellular Location: Nucleus
Initiation of transcription by RNA polymerase II requires the assistance of TATA box-binding protein (TBP) and TBP-associated factors, or TAFs (e.g., TAF2B), in 2 distinct complexes, TFIID and B-TFIID. The TFIID complex is composed of TBP and more than 8 TAFs. However, the majority of TBP is present in the B-TFIID complex, which is composed of TBP and TAFII170, also called TAF172, and has DNA-dependent ATPase activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent helicase BTAF1; B-TFIID transcription factor-associated 170 kDa subunit; BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, (Mot1 homolog, S. cerevisiae); BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1 homolog, S. cerevisiae); Mot1 homolog; TAF(II)170; TAF-172; TATA-binding protein-associated factor 172; TBP-associated factor 172
Gene Aliases: BTAF1; MOT1; RGD1564130; TAF(II)170; TAF172; TAFII170
UniProt ID: (Human) B4E0W6
Entrez Gene ID: (Human) 9044, (Rat) 368042
Molecular Function: DNA helicase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support