Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: TKSGSTPLP LSGGTSSSDN SWTMDASIDE VVRKAKLGTP SNMPPNFPYI SSPSVSPPTP EPLHKVYEEK TKLPSSVEVK KKLKKELKTK MKKKEKQRDR EREKDKNKDK SKEKDKVKEK EKDKETGRET KYPWKEFLKE EEADPYKFKI KEFEDVDPKV KLKDGLVRKE KEKHKDKKKD REKGKKDKDK REKEKVKDKG REDKMKAPAP PLVLPPKELA LPLFSPATAS RVPAMLPSLL PVLPEKLFEE KEKVKEKEKK KDKKEKKKKK EKEKEKKEK (443-721 aa encoded by BC024661)
Transcription factor TFIID is one of the general factors required for accurate and regulated initiation by RNA polymerase II. TFIID is a multimeric protein complex that plays a central role in mediating promoter responses to various activators and repressors. Required in complex with TBPL2 for the differentiation of myoblasts into myocytes. The complex replaces TFIID at specific promoters at an early stage in the differentiation process. Belongs to the TFIID complex which is composed of TATA binding protein (TBP) and a number of TBP-associated factors (TAFs). Interacts with TAF10 via the histone fold. Interacts with TAF13, TBP, SAP130 and GCN5L2. Interacts with TBPL2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 140 kDa TATA box-binding protein-associated factor; MGC25063; RNA polymerase II transcription factor TAFII140; TAF(II)140; TAF3 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 140kDa; TATA box binding protein associated factor 3; TBP-associated factor 3; Transcription initiation factor TFIID 140 kDa subunit; Transcription initiation factor TFIID subunit 3
Gene Aliases: TAF140; TAF3; TAFII-140; TAFII140
UniProt ID: (Human) Q5VWG9
Entrez Gene ID: (Human) 83860
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support