Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Immunogen sequence: PHLVPFLKKS VVALRQLLPN SQSFIQQCVQ QTSSDMVIAT CTTTVTTSPV VTTTVSSSQS EKSIIVSGAT APRTVSVQTL NPLAGPVGAK AGVVTLHSVG PTAATGGTTA GTGLLQTSKP LVTSVANTVT TVSLQPEKPV VSGTAVT
Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 70%.
TAF II p105, also called TAF4B, is a celltype specific transcriptional co-activator that is a component of the TFIID complex. Expressed primarily in B cells and ovarian granulosa cells, TAF II p105 can interact with OCBA/POU2AF1 to activate B cell-specific octamerdependent transcription. Additionally, TAF II p105 plays an important role in co-activating the transcription factor NFkB and is essential for activation of anti-apoptotic genes such as TNFAIP3. TAF II p135, also known as TAF4, TAF2C, TAF2C1, TAF4A or TAFII130, is a 1,085 amino acid subunit of TFIID that accelerates transcriptional activation triggered by thyroid hormone (TR) or retinoic acid (RA). Localized to the nucleus, TAF II p135 contains one TAFH domain and is thought to bind to proteins that contain glutamine-rich domains, such as the transcription factor CREB.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TAF(II)105; TAF4b RNA polymerase II, TATA box binding protein (TBP)-associated factor, 105kDa; TAFII-105; TATA box binding protein (TBP)-associated factor 4B; TATA box binding protein (TBP)-associated factor, RNA polymerase II, C2, 105kD; TATA box binding protein associated factor 4b; Transcription initiation factor TFIID 105 kDa subunit; Transcription initiation factor TFIID subunit 4B
Gene Aliases: SPGF13; TAF2C2; TAF4B; TAFII105
UniProt ID: (Human) Q92750
Entrez Gene ID: (Human) 6875
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support