Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLLPNILLT GTPGVGKTTL GKELASKSGL KYINVGDLAR EEQLYDGYDE EYDCPILDED RVVDELDNQM REGGVIVDYH GCDFFPERWF HIVFVLRTDT NVLYERLETR GYNEKKLTDN IQCEIFQVLY EEATASYKEE IVHQLPSNKP EELENNVDQI LKWIEQWIKD HNS (1-172 aa encoded by BC007426)
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the smaller subunits of TFIID that binds to the basal transcription factor GTF2B as well as to several transcriptional activators such as p53 and VP16. A similar but distinct gene (TAF9L) has been found on the X chromosome and a pseudogene has been identified on chromosome 19. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AD-004; CGI-137; CIP; hCINAP; MGC:1603; MGC:3647; RNA polymerase II TBP-associated factor subunit G; STAF31/32; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32 kDa; TAF9 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 32kDa; TAF9 RNA polymerase II, TATA box binding protein-associated factor; taf9 {ECO:0000312|EMBL:AAH91109.1}; TAFII-31; TAFII-32; TATA box binding protein associated factor 9; transcription initiation factor TFIID 31 kD subunit; Transcription initiation factor TFIID 31 kDa subunit; Transcription initiation factor TFIID 32 kDa subunit; Transcription initiation factor TFIID subunit 9
Gene Aliases: 2310012M09Rik; AA673500; AU045423; CINAP; MGC:5067; STAF31/32; TAF2G; TAF9; TAFII-31; TAFII-32; TAFII31; TAFII32; TAFIID32
UniProt ID: (Human) Q16594, (Rat) Q5BKE0, (Mouse) Q8VI33
Entrez Gene ID: (Human) 6880, (Rat) 373541, (Mouse) 108143
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support