Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA1, 2, 4, or 5 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chemokine-like protein TAFA-3; family with sequence similarity 19 (chemokine (C-C motif)-like), member A3
Gene Aliases: FAM19A3; TAFA-3; TAFA3
UniProt ID: (Human) Q7Z5A8
Entrez Gene ID: (Human) 284467
Molecular Function: chemokine
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support