NeoBiotechnologies
FIGURE: 1 / 2
Immunohistochemistry (PFA fixed): incubate antibody for 30 min RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.
Cancer Antigen 72-4 (CA 72-4, TAG 72-4) is a tumor marker that may be used to diagnose and monitor neoplastic diseases.
Protein Aliases: CA72-4; TAG-72