Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKLRSSHNAS KTLNANNMET LIECQSEGDI KEHPLLASCE SEDSICQLIE VKKRKKVLSW PFLMRRLSPA SDFSGALETD LKASLFDQPL SIICGDSDTL PRPIQDILTI LCLKGPSTEG IFRRAANEKA RKELKEELNS GDAVDLERLP VHLLAVVFKD FLRSIPRKLL SSDLFEEWMG ALEMQDEEDR IEALKQVADK LPRPNLLLLK HLVYVLHLIS KNSEVNRMDS SNLAICIGPN MLTLENDQSL SFEAQKDLNN KVKTLVEFLI DNCFEIFGEN IPVHSSITSD DSLEHTDSSD VSTLQNDSAY DSNDPDVESN SSSGISSPSR QPQVPMATAA GLDSAGPQDA REVSPEPIVS TVARLKSSLA QPDRRYSEPS MPSSQECLES RVTNQTLTKS EGDFPVPRVG SRLESEEAED PFPEEVFPAV QGKTKRPVDL KIKNLAPGSV LPRALVLKAF SSSSLDASSD SSPVASPSSP KRNFFSRHQS FTTKTEKGKP SREIKKHSMS FTFAPHKKVL TKNLSAGSGK SQDFTRDHVP RGVRKESQLA GRIVQENGCE THNQTARGFC LRPHALSVDD VFQGADWERP GSPPSYEEAM QGPAARLVAS ESQTVGSMTV GSMRARMLEA HCLLPPLPPA HHVEDSRHRG SKEPLPGHGL SPLPERWKQS RTVHASGDSL GHVSGPGRPE LLPLRTVSES VQRNKRDCLV RRCSQPVFEA DQFQYAKESY I
TAGAP (T-cell activation Rho GTPase-activating protein) is a 731 amino acid protein that contains one Rho-GAP domain and exists as 4 alternatively spliced isoforms. TAGAP may function as a GTPase-activating protein and may play important roles during T-cell activation. The gene that encodes TAGAP contains approximately 10,685 bases and maps to human chromosome 6q25.3. Making up nearly 6% of the human genome, chromosome 6 contains around 1,200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins, are also located on chromosome 6. In addition, Stickler syndrome, hydroxylase deficiency and maple syrup urine disease are associated with genes on chromosome 6, while a bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T-cell activation GTPase-activating protein; T-cell activation Rho GTPase-activating protein
Gene Aliases: ARHGAP47; FKSG15; IDDM21; TAGAP; TAGAP1
UniProt ID: (Human) Q8N103
Entrez Gene ID: (Human) 117289
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support