Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTERPPSEAA RSDPQLEGRD AAEASMAPPH LVLLNGVAKE TSRAAAAEPP VIELGARGGP GGGPAGGGGA ARDLKGRDAA TAEARHRVPT TELCRPPGPA PAPAPASVTA ELPGDGRMVQ LSPPALAAPA APGRALLYSL SQPLASLGSG FFGEPDAFPM FTTNNRVKRR PSPYEMEITD GPHTKVVRRI FTNSRERWRQ QNVNGAFAEL RKLIPTHPPD KKLSKNEILR LAMKYINFLA KLLNDQEEEG TQRAKTGKDP VVGAGGGGGG GGGGAPPDDL LQDVLSPNSS CGSSLDGAAS PDSYTEEPAP KHTARSLHPA MLPAADGAGP R
TAL-1 is a basic helix-loop-helix transcription. It regulates differentiation and survival during hemopoiesis. TAL-1 is also implicated in the genesis of hemopoietic malignancies and it may play an important role in hemopoietic differentiation. It serves as a positive regulator of eryhtroid differentiation. Mutations are associated with T-cell leukemia and melanoma. TAL-1 is know to bind to the LIM domain containing protein Rhombotin-2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa17; Class A basic helix-loop-helix protein 17; HGNC:11556; Stem cell protein; T-cell acute lymphocytic leukemia 1; T-cell acute lymphocytic leukemia protein 1; T-cell leukemia/lymphoma protein 5; TAL-1; tal-1 product
Gene Aliases: BHLHA17; SCL; tal-1; TAL1; TCL5
UniProt ID: (Human) P17542
Entrez Gene ID: (Human) 6886
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support