Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This product was prepared from monospecific antiserum by delipidation, salt fractionation and ion exchange chromatography. A single precipitin arc was observed against anti-Rabbit Serum when assayed by immunoelectrophoresis.
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is involved in the pumping of degraded cytosolic peptides across the endoplasmic reticulum into the membrane-bound compartment where class I molecules assemble. Mutations in this gene may be associated with ankylosing spondylitis, insulin-dependent diabetes mellitus, and celiac disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABC transporter, MHC 1; Antigen peptide transporter 1; AOI; APT1; ATP-binding cassette sub-family B member 2; ATP-binding cassette, sub-family B (MDR/TAP), member 2; beta-filamin; DAAP-57C1.5; FH1; filamin homolog 1; filamin-3; filamin-B; FLJ26666; FLJ41500; FLN1L; LRS1; Peptide supply factor 1; Peptide transporter involved in antigen processing 1; Peptide transporter PSF1; Peptide transporter TAP1; PSF-1; Really interesting new gene 4 protein; RING4; TABP; tap i; thyroid autoantigen; transporter 1 ATP-binding cassette sub-family B; transporter 1, ATP-binding cassette, sub-family B (MDR/TAP); transporter associated with antigen processing; transporter, ATP-binding cassette, major histocompatibility complex, 1
Gene Aliases: ABC17; ABCB2; APT1; D6S114E; PSF-1; PSF1; RING4; TAP1; TAP1*0102N; TAP1N; Y3
UniProt ID: (Human) Q03518
Entrez Gene ID: (Human) 6890
Molecular Function: ATP-binding cassette (ABC) transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support