Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Sequence of this protein is as follows: MPLHVKWPFP AVPPLTWTLA SSVVMGLVGT YSCFWTSEWA QAEAGPPGYP CPAGGILKLR HIWNLKLMRW TPAAADICFT KELHSHFFSL GKCVPVCRGD GVYQKGMDFI LEKLNHGDWV HIFPEGIGRL IAECHLNPII LPLWHVGEPG DGDREMASGV GGLGLPLVPG CPAPPHVWPS VHCAAGMNDV LPNSPPYFPR FGQKITVLIG KPFSALPVLE RLRAENKSAV EMRKALTDFI QEEFQHLKTQ AEQLHNHLQP GR
This gene encodes a protein that is expressed at high levels in cardiac and skeletal muscle. Mutations in this gene have been associated with a number of clinical disorders including Barth syndrome, dilated cardiomyopathy, hypertrophic DCM, endocardial fibroelastosis, and left ventricular noncompaction. Multiple transcript variants encoding different isoforms have been described. A long form and a short form of each of these isoforms is produced; the short form lacks a hydrophobic leader sequence and may exist as a cytoplasmic protein rather than being membrane-bound. Other alternatively spliced transcripts have been described but the full-length nature of all these transcripts is not known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ27390; HGNC:11577; OTTHUMP00000061673; Protein G4.5; Tafazzin; Taz; XAP-2; XX-FW83563B9.3
Gene Aliases: BTHS; CMD3A; EFE; EFE2; G4.5; LVNCX; TAFAZZIN; TAZ; Taz1
UniProt ID: (Human) Q16635
Entrez Gene ID: (Human) 6901
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support