Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LGFSDIHFNS RNTFQVSRGQ SARDHLPPAG PPVPLPAAEQ GPAGASARAR RSGGFADFFT RNLFPKRTKE LKSVVHSAPG WKLFGKVPPR ENLQKTSKII QQEYEART
Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TBC1 domain family member 12; TBC1 domain family, member 12
Gene Aliases: KIAA0608; TBC1D12
UniProt ID: (Human) O60347
Entrez Gene ID: (Human) 23232
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support