Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MALSDEPAA GGPEEEAEDE TLAFGAALEA FGESAETRAL LGRLREVHGG GAEREVALER FRVIMDKYQE QPHLLDPHLE WMMNLLLDIV QDQTSPASLV HLAFKFLYII TKVRGYKTFL RLFPHEVADV EPVLDLVTIQ NPKDHEAWET RYMLLLWLSV TCLIPFDFSR LDGNLLTQPG QARMSIMDRI LQIAESYLIV SDKARDAAAV LVSRFITRPD VKQSKMAEFL DWSLCNLARS SFQTMQGVIT MDGTLQALAQ IFKHGKREDC L (1-270 aa encoded by BC003094)
Cofactor D is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-tubulin cofactor D; SSD-1; tfcD; Tubulin-folding cofactor D; Tubulin-specific chaperone D
Gene Aliases: 2310057L06Rik; A030005L14Rik; KIAA0988; mKIAA0988; PP1096; SSD-1; SSD1; TBCD; TFCD
UniProt ID: (Human) Q9BTW9, (Mouse) Q8BYA0
Entrez Gene ID: (Human) 6904, (Rat) 100361417, (Mouse) 108903
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support