Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
This gene is a member of a conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similar protein has been disrupted in mice and shown to be critical for early cortical development, and causes loss of projection neurons in the olfactory bulbs and olfactory cortex. The C-terminal region this similar protein was found to be necessary and sufficient for association with the guanylate kinase domain of calcium/calmodulin-dependent serine protein kinase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T-box brain gene 1; T-box brain protein 1; T-box, brain 1; T-box, brain, 1; T-box, brain, 1-like; T-brain-1; TBR-1; TES-56
Gene Aliases: TBR-1; TBR1; TES-56
UniProt ID: (Human) Q16650, (Mouse) Q64336
Entrez Gene ID: (Human) 10716, (Pig) 100154282, (Mouse) 21375, (Rat) 680427
Molecular Function:
Rel homology transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support