Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEFTASPKPQ LSSRANAFSI AALMSSGGSK EKEATENTIK PLEQFVEKSS CAQPLGELTS LDAHGEFGGG SGSSPSSSSL CTEPLIPTTP IIPSEEMAKI ACSLETKELW DKFHELGTEM IITKSGRRMF PTIRVSFSGV DPEAKYIVLM DIVPVDNKRY RYAYHRSSWL VAGKADPPLP ARLYVHPDSP FTGEQLLKQM VSFEKVKLTN NELDQHGHII LNSMHKYQPR VHIIKKKDHT ASLLNLKSEE FRTFIFPETV FTAVTAYQNQ LITKLKIDSN PFAKGFRDSS RLTDIER
TBX20 is a member of the T-box transcription factor family expressed in the developing heart, eye, ventral neural tube, and limbs, indicating a possible role in regulating development of these tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T-box protein 20; T-box transcription factor TBX20
Gene Aliases: ASD4; TBX20
UniProt ID: (Human) Q9UMR3
Entrez Gene ID: (Human) 57057
Molecular Function: Rel homology transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support