Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: MEKLFNENEG MPSNQGKIDN EEQPPHEGKP EVACILEDKK LENEGNTENT
Sequence homology: Human: 100%
TCEAL2 is a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner.This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family contain TFA domains and may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. Multiple family members are located on the X chromosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TCEA-like protein 2; transcription elongation factor A (SII)-like 2; Transcription elongation factor A protein-like 2; Transcription elongation factor S-II protein-like 2
Gene Aliases: My048; MY0876G0; MY0876G05; TCEAL2; WEX1
UniProt ID: (Human) Q9H3H9
Entrez Gene ID: (Human) 140597
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support