Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAAESALQVV EKLQARLAAN PDPKKLLKYL KKLSTLPITV DILAETGVGK TVNSLRKHEH VGSFARDLVA QWKKLVPVER NAEPDEQDFE KSNSRKRPRD ALQKEEEMEG DYQETWKATG SRSYSPDHRQ KKHRKLSELE RPHKVSHGHE RRDERKRCHR MSPTYSSDPE SSDYGHVQSP PSCTSPHQMY VDHYRSLEED QEPIVSHQKP GKGHSNAFQD RLGASQERHL GEPHGKGVVS QNKEHKSSHK DKRPVDAKSD EKASVVSREK SHKALSKEEN RRPPSGDNAR EKPPSSGVKK EKDREGSSLK KKCLPPSEAA SDNHLKKPKH RDPEKAKLDK SKQGLDSFDT GKGAGDLLPK VKEKGSNNLK TPEGKVKTNL DRKSLGSLPK VEETDMEDEF EQPTMSFESY LSYDQPRKKK KKIVKTSATA LGDKGLKKND SKSTGKNLDS VQKLPKVNKT KSEKPAGADL AKLRKVPDVL PVLPDLPLPA IQANYRPLPS LELISSFQPK RKAFSSPQEE EEAGFTGRRM NSKMQVYSGS KCAYLPKMMT LHQQCIRVLK NNIDSIFEVG GVPYSVLEPV LERCTPDQLY RIEEYNHVLI EETDQLWKVH CHRDFKEERP EEYESWREMY LRLQDAREQR LRVLTKNIQF AHANKPKGRQ AKMAFVNSVA KPPRDVRRRQ EKFGTGGAAV PEKIKIKPAP YPMGSSHASA SSISFNPSPE EPAYDGPSTS SAHLAPVVSS TVSYDPRKPT VKKIAPMMAK TIKAFKNRFS RR
This gene encodes the transcriptionally active subunit of the SIII (or elongin) transcription elongation factor complex, which also includes two regulatory subunits, elongins B and C. This complex acts to increase the rate of RNA chain elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites along the DNA template. Whereas a related protein with similar function, elongin A, is ubiquitously expressed, the encoded protein is specifically expressed in the testis, suggesting it may have a role in spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EloA; EloA2; Elongin 110 kDa subunit; elongin A; Elongin A2; Elongin-A; Elongin-A2; RNA polymerase II transcription factor SIII subunit A1; RNA polymerase II transcription factor SIII subunit A2; SIII p110; TCEB3B; TCEB3L; transcription elongation factor (SIII) elongin A2 elongin A2; transcription elongation factor B (SIII), polypeptide 3 (110kDa, elongin A); transcription elongation factor B alpha subunit; Transcription elongation factor B polypeptide 3; Transcription elongation factor B polypeptide 3B
Gene Aliases: ELOA; ELOA2; HsT832; MSTP059; SIII; SIII p110; TCEB3; TCEB3A; TCEB3B; TCEB3L
UniProt ID: (Human) Q14241, (Human) Q8IYF1
Entrez Gene ID: (Human) 6924, (Human) 51224
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support