Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: SEGSSQPEEQ LKSPMAESLD GGCSSSSEDQ GERVRQLSGQ STSSDTTYKG GASEKAGSSP AQGAQNEPPR LNASPAAREE ATSPGAKDMP LSSDGNPKVN EKTVGVIVS
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
This gene encodes a transcription factor that recognizes the platelet-derived growth factor-responsive element in the matrix metalloproteinase 3 promoter. The encoded protein is thought to be a transcriptional coactivator, enhancing the activity of transcription factors such as JUN and SP1. Mutations in this gene are associated with autism spectrum disorders. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear factor SPBP; Protein AR1; SPRE-binding protein; Stromelysin-1 PDGF-responsive element-binding protein; stromelysin-1 platelet-derived growth factor-responsive element binding protein; TCF-20; Transcription factor 20; transcription factor 20 (AR1)
Gene Aliases: AR1; KIAA0292; SPBP; TCF-20; TCF20
UniProt ID: (Human) Q9UGU0
Entrez Gene ID: (Human) 6942
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support