Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
This antibody detects endogenous protein at a molecular weight of 60 and 72 kDa.
Purity is >95% by SDS-PAGE.
T cell factors (TCFs) comprise a family of DNA-binding transcriptional activators that are essential for lymphoid cell development. These transcription factors are activated by the Wnt-1 and Wingless pathways and are characterized by the presence of a conserved protein motif, the high mobility group(HMG) 1 box, which mediates DNA binding. TCF-4 mainly localizes in the cytoplasm and is transported into the nucleus directly bound to beta-catenin in a cooperative manner. This TCF-4/ beta-catenin complex induces expression of Wnt target genes, including multiple cancer-associated genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHb19; bHLHb20; Class A helix-loop-helix transcription factor ME1; Class A helix-loop-helix transcription factor ME2; Class B basic helix-loop-helix protein 19; Class B basic helix-loop-helix protein 20; DNA-binding protein HTF4; E-box-binding protein; helix-loop-helix transcription factor 4; Immunoglobulin transcription factor 2; immunoglobulin transcription factor-2; ITF-2; MITF-2; R8f DNA-binding protein; RITF-2; Salivary-specific cAMP response element-binding protein alpha; SCBP alpha; SCBP-alpha; SEF-2; SL3-3 enhancer factor 2; TCF-12; TCF-4; Transcription factor 12; Transcription factor 4; Transcription factor 4 (Immunoglobulin transcription factor 2) (ITF-2) (MITF-2) (SL3-3 enhancer factor 2) (SEF-2) (Class A helix-loop-helix transcription factor ME2); transcription factor 4, isoform C; transcription factor 4, isoform D; transcription factor 4, isoform E; transcription factor 4, isoform L; transcription factor 4, isoform M; transcription factor 4, isoform R; Transcription factor HTF-4
Gene Aliases: 5730422P05Rik; A130037E08Rik; ALF1; ASP-I2; BHLHB19; BHLHB20; CRS3; E2-2; E2.2; FECD3; HEB; HEBAlt; HsT17266; HTF-4; HTF4; ITF-2; ITF-2b; ITF2; ME1; ME2; Meb; MITF-2A; MITF-2B; PTHS; REB; SCBPA; SEF-2; SEF2; SEF2-1; SEF2-1A; SEF2-1B; SEF2-1D; TCF-12; TCF-4; TCF12; TCF4; TFE
UniProt ID: (Human) Q99081, (Human) P15884, (Mouse) Q61286, (Rat) P51514, (Mouse) Q60722, (Rat) Q62655
Entrez Gene ID: (Human) 6938, (Human) 6925, (Mouse) 21406, (Rat) 25720, (Mouse) 21413, (Rat) 84382
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support