Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence:
GAVGQIPHP LGWLVPQQGQ PMYSLPPGGF RHPYPALAMN ASMSSLVSSR FSPHMVAPAH PGLPTSGIPH PAIVSPIVKQ EPAPPSLSPA VSVKSPVTVK KEEEKKPHVK KPLNAFMLYM KEMRAKVVAE CTLKESAAIN QILGRKWHNL SREEQAKYYE LARKERQLHS QLYPTWSARD NYGKKKKRKR EKQLSQTQSQ QQVQEAEGAL ASKSKKPCVQ YLPPEKPCDS PASSHGSMLD SPATPSAALA SPAAPAATHS EQAQPLSLTT KPETRAQLAL HSAAFLSAKA AASSSRQMGS QPPLLSRPLP LGSMPTALLA SPPSFPATLH AHQALPVLQA QPLSLVTKSA H
This gene encodes a member of the T cell factor/lymphoid enhancer factor family of transcription factors. These transcription factors are activated by beta catenin, mediate the Wnt signaling pathway and are antagonized by the transforming growth factor beta signaling pathway. The encoded protein contains a high mobility group-box DNA binding domain and participates in the regulation of cell cycle genes and cellular senescence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HMG box transcription factor 3; T-cell factor 3; TCF-3; transcription factor 3; Transcription factor 7-like 1; transcription factor 7-like 1 (T-cell specific, HMG box); transcription factor 7-like 1 (T-cell specific, HMG-box); transcription factor 7-like 2 (T-cell specific, HMG box)
Gene Aliases: bHLHb21; TCF-3; TCF3; TCF7L1
UniProt ID: (Human) Q9HCS4, (Mouse) Q9Z1J1
Entrez Gene ID: (Human) 83439, (Mouse) 21415
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support