Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockdown to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 6
It is not recommended to aliquot this antibody.
TCF7L2, or TCF4, is a high mobility group (HMG) box-containing transcription factor that plays a key role in the Wnt signaling pathway. TCF7L2 has been implicated in glucose homeostasis through the regulation of pro-glucagon gene expression, which encodes glucagon-like peptide 1 (GLP-1). Genetic variants of TCF7L2 have been linked to type 2 diabetes, and it has specifically been shown to have an important role in the regulation of both beta-cell survival and function. Several splicing isoforms for TCF7L2 have been shown to be differentially expressed in tissues during cancer progression, and gene targeting studies indicate that TCF7L2 is required to maintain the crypt stem cells of the small intestine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HMG box transcription factor 4; hTCF; hTCF 4; hTCF-4; RP11-357H24.1; T-cell factor 4; T-cell factor-4 variant A; T-cell factor-4 variant B; T-cell factor-4 variant C; T-cell factor-4 variant D; T-cell factor-4 variant E; T-cell factor-4 variant F; T-cell factor-4 variant G; T-cell factor-4 variant H; T-cell factor-4 variant I; T-cell factor-4 variant J; T-cell factor-4 variant K; T-cell factor-4 variant L; T-cell factor-4 variant M; T-cell factor-4 variant X2; T-cell-specific transcription factor 4; TCF 4; TCF7L2 protein; Transcription factor 7-like 2; transcription factor 7-like 2 (T-cell specific, HMG-box)
Gene Aliases: TCF-4; TCF4; TCF7L2
UniProt ID: (Human) Q9NQB0
Entrez Gene ID: (Human) 6934
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support