Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
TCFL5 is a new bHLH transcription factor that negatively regulates upstream transcription factor-dependent transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cha transcription factor; E2BP-1; HPV-16 E2-binding protein 1; MGC46135; transcription factor-like 5 (basic helix-loop-helix); Transcription factor-like 5 protein
Gene Aliases: bHLHe82; CHA; E2BP-1; E2BP1; Figlb; TCFL5
UniProt ID: (Human) Q9UL49
Entrez Gene ID: (Human) 10732
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support