Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MALPTLPSY WCSQQRLNQQ LARQREQEAR LRQQWEQNSR YFRMSDICSS KQAEWSSKTS YQRSMHAYQR EKMKEEKRRS LEARREKLRQ LMQEEQDLLA RELEELRLSM NLQERRIREQ HGKLKSAKEE QRKLIAEQLL YEHWKKNNPK LREMELDLHQ KHVVNSWEMQ KEEKKQQEAT AEQENKRYEN EYERARREAL ERMKAEEERR QLEDKLQAEA LLQQMEELKL KEVEATKLKK EQENLLKQRW ELERLEEERK QMEAFRQKAE LGRFLRHQYN AQLSRRTQQI QEELEADRRI LQALLE (1-305 aa encoded by B C004285)
TCHP may act as a 'capping' or 'branching' protein for keratin filaments in the cell periphery. This protein may regulate K8/K18 filament and desmosome organization mainly at the apical or peripheral regions of simple epithelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial protein with oncostatic activity; Mitostatin; Protein TCHP; TCHP; Trichoplein keratin filament-binding protein; trichoplein, keratin filament binding; Tumor suppressor protein
Gene Aliases: TCHP; TpMs
UniProt ID: (Human) Q9BT92
Entrez Gene ID: (Human) 84260
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support