Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: CVSRLGELGL VEFRDLNASV SAFQRRFVVD VRRCEELEKT FTFLQEEVRR AGLVLPPPKG RLPAPPPRDL LRIQEETERL AQELRDVRGN QQALRAQLHQ LQLHAAVLRQ GHEPQLAAAH TDGASERTPL LQAPGGPHQD LRVNFVAGAV EPHKAPALER LLWRACRGFL IASFRELEQP LEHPVTGEPA TWMTFLISYW GEQIGQKIRK ITDCFHCHVF PFLQQEEARL GALQQLQQQS QELQEVLGET ERFLSQVLGR VLQLLPPGQV QVHKMKAVYL ALNQCSVSTT HKCLIAEAWC SVRDLPALQE ALRDSSMEEG VSAVAHRIPC RDMPPTLIRT NRFTASFQGI VDAYGVGRYQ EVNPAPYTII T
The T-cell immune regulator 1 (TCIRG1) is the a3 subunit of the vacuolar-type proton transporting ATPase (V-ATPase), a proton pump in late endosomes and lysosomes that functions in the luminal acidification of these organelles. Genetic defects in the gene encoding this protein a responsible for a severe form of autosomal recessive osteopetrosis, a condition that leads to increased bone density, decreased bone strength, and inflammation in bone tissues. TCIRG1 is also required for the normal secretion of insulin and the bacteria-killing function of macrophages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, H+ transporting, 116kD; OC-116 kDa; Osteoclastic proton pump 116 kDa subunit; specific 116-kDa vacuolar proton pump subunit; T-cell immune regulator 1; T-cell immune response cDNA 7; T-cell immune response cDNA7 protein; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 protein a; T-cell, immune regulator 1, ATPase, H+ transporting, lysosomal V0 subunit A3; TIRC7; V-ATPase 116 kDa subunit a 3; V-ATPase 116-kDa; V-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a 3; vacuolar proton translocating ATPase 116 kDa subunit A; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 3
Gene Aliases: a3; Atp6i; ATP6N1C; ATP6V0A3; OC-116kDa; OC116; OPTB1; Stv1; TCIRG1; TIRC7; Vph1
UniProt ID: (Human) Q13488
Entrez Gene ID: (Human) 10312
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support