Product References
Continuous Antigenic Stimulation of DO11.10 TCR Transgenic Mice in the Presence or Absence of IL-1β: Possible Implications for Mechanisms of T Cell Depletion in HIV Disease.
Journal of immunology (Baltimore, Md. : 1950)
Ladell K,Hazenberg MD,Fitch M,Emson C,McEvoy-Hein Asgarian BK,Mold JE,Miller C,Busch R,Price DA,Hellerstein MK,McCune JM
MM7505 was used in flow cytometry to explore the impact of chronic antigenic stimulation on CD4(+) T cell dynamics.
Sun Nov 01 00:00:00 EDT 2015
Accumulation of effector CD4 T cells during type 2 immune responses is negatively regulated by Stat6.
Journal of immunology (Baltimore, Md. : 1950)
King SB,Knorn AM,Ohnmacht C,Voehringer D
MM7505 was used in flow cytometry to investigate the effect of Stat6 in T cell expansion, survival, and recruitment to the lung
Tue Jan 15 00:00:00 EST 2008
Influenza A virus elevates active cathepsin B in primary murine DC.
International immunology
Burster T,Giffon T,Dahl ME,Björck P,Bogyo M,Weber E,Mahmood K,Lewis DB,Mellins ED
MM7505 was used in flow cytometry to suggest that the influenza A virus affects the MHC class II antigen-processing pathway
Tue May 01 00:00:00 EDT 2007
Structure and duration of contact between dendritic cells and T cells are controlled by T cell activation state.
European journal of immunology
Rothoeft T,Balkow S,Krummen M,Beissert S,Varga G,Loser K,Oberbanscheidt P,van den Boom F,Grabbe S
MM7505 was used in flow cytometry to characterize T cells interactions with dendritic cells
Fri Dec 01 00:00:00 EST 2006
CD25+Foxp3+ regulatory T cells facilitate CD4+ T cell clonal anergy induction during the recovery from lymphopenia.
Journal of immunology (Baltimore, Md. : 1950)
Vanasek TL,Nandiwada SL,Jenkins MK,Mueller DL
MM7505 was used in flow cytometry to study ineffective T cell clonal anergy induction in nu/nu mice
Mon May 15 00:00:00 EDT 2006
Divergent generation of heterogeneous memory CD4 T cells.
Journal of immunology (Baltimore, Md. : 1950)
Moulton VR,Bushar ND,Leeser DB,Patke DS,Farber DL
MM7505 was used in flow cytometry to elucidate the generation of memory CD4 T cells
Sat Jul 15 00:00:00 EDT 2006
Balanced expression of CXCR5 and CCR7 on follicular T helper cells determines their transient positioning to lymph node follicles and is essential for efficient B-cell help.
Blood
Hardtke S,Ohl L,Förster R
MM7505 was used in flow cytometry to report that CCR7 and CXCR5 determine the positioning and proper function of follicular T helper cells
Thu Sep 15 00:00:00 EDT 2005
The ratio between dendritic cells and T cells determines the outcome of their encounter: proliferation versus deletion.
European journal of immunology
Höpken UE,Lehmann I,Droese J,Lipp M,Schüler T,Rehm A
MM7505 was used in flow cytometry to study how partially mature dendritic cells regulate the induction of antigen-specific T cell proliferation and T cell tolerance
Sat Oct 01 00:00:00 EDT 2005
Increased Fas ligand expression of CD4+ T cells by HCV core induces T cell-dependent hepatic inflammation.
Journal of leukocyte biology
Cruise MW,Melief HM,Lukens J,Soguero C,Hahn YS
MM7505 was used in flow cytometry to elucidate the role of hepatitis C virus core-expressing T cells in liver inflammation
Mon Aug 01 00:00:00 EDT 2005
A role for CD28 in lymphopenia-induced proliferation of CD4 T cells.
Journal of immunology (Baltimore, Md. : 1950)
Hagen KA,Moses CT,Drasler EF,Podetz-Pedersen KM,Jameson SC,Khoruts A
MM7505 was used in flow cytometry to characterize T cells during lymphopenia-induced proliferation
Wed Sep 15 00:00:00 EDT 2004
Indirect minor histocompatibility antigen presentation by allograft recipient cells in the draining lymph node leads to the activation and clonal expansion of CD4+ T cells that cause obliterative airways disease.
Journal of immunology (Baltimore, Md. : 1950)
Richards DM,Dalheimer SL,Ehst BD,Vanasek TL,Jenkins MK,Hertz MI,Mueller DL
MM7505 was used in flow cytometry to investigate minor histocompatibility antigen-induced chronic allograft rejection.
Mon Mar 15 00:00:00 EST 2004
Vitamin A deficiency increases the in vivo development of IL-10-positive Th2 cells and decreases development of Th1 cells in mice.
The Journal of nutrition
Stephensen CB,Jiang X,Freytag T
MM7505 was used in flow cytometry to test if vitamin A deficiency affects development of Th1/Th2 memory cell phenotype in vivo
Fri Oct 01 00:00:00 EDT 2004
A limited antigen-specific cellular response is sufficient for the early control of Mycobacterium tuberculosis in the lung but is insufficient for long-term survival.
Infection and immunity
Turner J,Dobos KM,Keen MA,Frank AA,Ehlers S,Orme IM,Belisle JT,Cooper AM
MM7505 was used in flow cytometry to identify Rv1411c as an important and potent inducer of protective T cells within the lungs of mice infected with Mycobacterium tuberculosis
Thu Jul 01 00:00:00 EDT 2004
Preferential accumulation of antigen-specific effector CD4 T cells at an antigen injection site involves CD62E-dependent migration but not local proliferation.
The Journal of experimental medicine
Reinhardt RL,Bullard DC,Weaver CT,Jenkins MK
MM7505 was used in flow cytometry to study the migration of antigen-specific T cells to nonlymphoid tissues.
Mon Mar 17 00:00:00 EST 2003