Product References
Phenotypic and functional characterization of pharmacologically expanded Vγ9Vδ2 T cells in pigtail macaques.
iScience
Barber-Axthelm IM,Wragg KM,Esterbauer R,Amarasena TH,Barber-Axthelm VRB,Wheatley AK,Gibbon AM,Kent SJ,Juno JA
TCR1732 was used in Flow Cytometry to show the tissue distribution and phenotype of in vivo pharmacologically expanded Vγ9Vδ2 T cells can be altered based on the antigen administration route, with implications for tissue trafficking and the clinical efficacy of Vγ9Vδ2 T cell immunotherapeutic.
Fri Mar 17 00:00:00 UTC 2023
γδ T Cells Contribute to Injury in the Developing Brain.
The American journal of pathology
Albertsson AM,Zhang X,Vontell R,Bi D,Bronson RT,Supramaniam V,Baburamani AA,Hua S,Nazmi A,Cardell S,Zhu C,Cantor H,Mallard C,Hagberg H,Leavenworth JW,Wang X
TCR1732 was used in Immunohistochemistry to highlight unique features of injury in the developing brain, where, unlike injury in the mature brain, γδT cells function as initiators of injury independently of common γδT-cell-associated cytokines.
Thu Mar 01 00:00:00 UTC 2018
HMBPP-deficient Listeria mutant immunization alters pulmonary/systemic responses, effector functions, and memory polarization of Vγ2Vδ2 T cells.
Journal of leukocyte biology
Frencher JT,Shen H,Yan L,Wilson JO,Freitag NE,Rizzo AN,Chen CY,Chen ZW
TCR1732 was used in flow cytometry to examine the effect of HMBPP-deficient Listeria mutant immunization
Mon Dec 01 00:00:00 UTC 2014
Characterization of γδT cells in naïve and HIV-infected chimpanzees and their responses to T-cell activators in vitro.
Journal of medical primatology
Hodara VL,Parodi LM,Chavez D,Smith LM,Lanford R,Giavedoni LD
TCR1732 was used in flow cytometry to study the in vitro response of gammadelta T-cell subsets from HIV1-infected and control chimpanzees to T-cell activators
Fri Aug 01 00:00:00 UTC 2014
Phosphoantigen/IL2 expansion and differentiation of Vγ2Vδ2 T cells increase resistance to tuberculosis in nonhuman primates.
PLoS pathogens
Chen CY,Yao S,Huang D,Wei H,Sicard H,Zeng G,Jomaa H,Larsen MH,Jacobs WR,Wang R,Letvin N,Shen Y,Qiu L,Shen L,Chen ZW
TCR1732 was used in flow cytometry to study the increased Tuberculosis resistance of non-human primates following phoshoantigen/IL-2 mediated expansion and differentiation of VgammaVdelta2 T-cells
Thu Aug 01 00:00:00 UTC 2013
Multieffector-functional immune responses of HMBPP-specific Vγ2Vδ2 T cells in nonhuman primates inoculated with Listeria monocytogenes ΔactA prfA*.
Journal of immunology (Baltimore, Md. : 1950)
Ryan-Payseur B,Frencher J,Shen L,Chen CY,Huang D,Chen ZW
TCR1732 was used in flow cytometry to study multi-effector functional Vgamma2Vdelta2 T cell responses against Listeria monocytogenes in nonhuman primates
Wed Aug 01 00:00:00 UTC 2012
Mechanisms underlying γδ T-cell subset perturbations in SIV-infected Asian rhesus macaques.
Blood
Harris LD,Klatt NR,Vinton C,Briant JA,Tabb B,Ladell K,Lifson J,Estes JD,Price DA,Hirsch VM,Brenchley JM
TCR1732 was used in flow cytometry to investigate T cells in chronically SIVsmE543-infected rhesus macaques
Thu Nov 18 00:00:00 UTC 2010
Gammadelta T cell immune manipulation during chronic phase of simian-human immunodeficiency virus infection [corrected] confers immunological benefits.
Journal of immunology (Baltimore, Md. : 1950)
Ali Z,Yan L,Plagman N,Reichenberg A,Hintz M,Jomaa H,Villinger F,Chen ZW
TCR1732 was used in Flow Cytometry to investigate whether a well-defined phosphoantigen-4-hydroxy-3-methyl-but-2-enyl pyrophosphate/IL-2 therapeutic regimen can overcome HIV-mediated immune suppression to massively expand polyfunctional Vgamma2Vdelta2 T cells.
Thu Oct 15 00:00:00 UTC 2009
Definition of APC presentation of phosphoantigen (E)-4-hydroxy-3-methyl-but-2-enyl pyrophosphate to Vgamma2Vdelta 2 TCR.
Journal of immunology (Baltimore, Md. : 1950)
Wei H,Huang D,Lai X,Chen M,Zhong W,Wang R,Chen ZW
TCR1732 was used in western blot to study the antigen presentation of a specific phosphoantigen
Wed Oct 01 00:00:00 UTC 2008
Human CD1-restricted T cell recognition of lipids from pollens.
The Journal of experimental medicine
Agea E,Russano A,Bistoni O,Mannucci R,Nicoletti I,Corazzi L,Postle AD,De Libero G,Porcelli SA,Spinozzi F
TCR1732 was used in flow cytometry to study the immune response of pollen lipids
Mon Jul 18 00:00:00 UTC 2005
In vivo immunomanipulation of V gamma 9V delta 2 T cells with a synthetic phosphoantigen in a preclinical nonhuman primate model.
Journal of immunology (Baltimore, Md. : 1950)
Sicard H,Ingoure S,Luciani B,Serraz C,Fournié JJ,Bonneville M,Tiollier J,Romagné F
TCR1732 was used in flow cytometry to investigate the effect of a synthetic agonist on the stimulation of Vgamma9Vdelta2 in monkeys
Sat Oct 15 00:00:00 UTC 2005
Drug-induced expansion and differentiation of V gamma 9V delta 2 T cells in vivo: the role of exogenous IL-2.
Journal of immunology (Baltimore, Md. : 1950)
Casetti R,Perretta G,Taglioni A,Mattei M,Colizzi V,Dieli F,D'Offizi G,Malkovsky M,Poccia F
TCR1732 was used in flow cytometry to report the functional characteristics of Vgamma9Vdelta2 T cells from Macaca fascicularis
Mon Aug 01 00:00:00 UTC 2005
Inhibition of adaptive Vgamma2Vdelta2+ T-cell responses during active mycobacterial coinfection of simian immunodeficiency virus SIVmac-infected monkeys.
Journal of virology
Zhou D,Lai X,Shen Y,Sehgal P,Shen L,Simon M,Qiu L,Huang D,Du GZ,Wang Q,Letvin NL,Chen ZW
TCR1732 was used in flow cytometry to investigate the effect of mycobacterial coinfection of simian immunodeficiency virus on adaptive Vgamma2Vdelta2+ T-cell responses
Sat Mar 01 00:00:00 UTC 2003