Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEAENAGSY SLQQAQAFYT FPFQQLMAEA PNMAVVNEQQ MPEEVPAPAP AQEPVQEAPK GRKRKPRTTE PKQPVEPKKP VESKKSGKSA KSKEKQEKIT DTFKVKRKVD RFNGVSEAEL LTKTLPDILT FNLDIVIIGI NPGLMAAYKG HHYPGPGNHF WKCLFMSGLS EVQLNHMDDH TLPGKYGIGF TNMVERTTPG SKDLSSKEFR EGGRILVQKL QKYQPRIAVF NGKCIYEIFS KEVFGVKVKN LEFGLQPHKI PDTETLCYGM PSSSARCAQF PRAQDKVHYY IKLKDLRDQL KGIERNMDVQ EVQYTFDLQL AQEDAKKMAV KEEKYDPGYE AAYGGAYGEN PCSSEPCGFS SNGLIESVEL RGESAFSGIP NGQWMTQSFT DQIPSFSNHC GTQEQEEESH A (1-410 aa encoded by BC037557)
The protein encoded by this gene belongs to the TDG/mug DNA glycosylase family. Thymine-DNA glycosylase removes thymine moieties from G/T mismatches by hydrolyzing the carbon-nitrogen bond between the sugar-phosphate backbone of DNA and the mis paired thymine. With lower activity, this enzyme also removes thymine from C/T and T/T mispairings. TDG can also remove uracil and 5-bromouracil from mispairings with guanine. This enzyme plays a central role in cellular defense against genetic mutation caused by the spontaneous deamination of 5-methylcytosine and cytosine. This gene may have a pseudogene in the p arm of chromosome 12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-JUN leucine zipper interactive protein JZA-3; G/T mismatch-specific thymine DNA glycosylase; hTDG; Thymine-DNA glycosylase
Gene Aliases: E130317C12Rik; hTDG; JZA-3; Jza1; TDG
UniProt ID: (Human) Q13569, (Mouse) P56581
Entrez Gene ID: (Human) 6996, (Rat) 114521, (Mouse) 21665
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support