Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDCRKMARFS YSVIWIMAIS KVFELGLVAG LGHQEFARPS RGYLAFRDDS IWPQEEPAIR PRSSQRVPPM GIQHSKELNR TCCLNGGTCM LGSFCACPPS FYGRNCEHDV RKENCGSVPH DTWLPKKCSL CKCWHGQLRC FPQAFLPGCD GLVMDEHLVA SRTPELPPSA RTTTFMLVGI CLSIQSYY
Cripto (TDGF1) is a GPI-anchored cell membrane proetin involved in Nodal signaling. Cell associated TDGF1 acts as a a Nodeal coreceptor in cis. Shedding of TDGF1 by TMEM8A modulated Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells. TDGF1 is expressed both in ES cells and during the early phases of embryo development, while in adults it is reactivated in a wide range of epithelial cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CRGF; Cripto, EGF-CFC family member; Cripto-1; Cripto-1 growth factor; Cripto-3; Epidermal growth factor-like cripto protein CR1; Protein Cripto; TDGF2; Teratocarcinoma-derived growth factor 1
Gene Aliases: CR; CRGF; CRIPTO; CRIPTO-1; TDGF1
UniProt ID: (Human) P13385
Entrez Gene ID: (Human) 6997
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support