Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MSQEGDYGR WTISSSDESE EEKPKPDKPS TSSLLCARQG AANEPRYTCS EAQKAAHKRK ISPVKFSNTD SVLPPKRQKS GSQEDLGWCL SSSDDELQPE MPQKQAEKVV IKKEKDISAP NDGTAQRTEN HGAPACHRLK EEEDEYETSG EGQDIWDMLD KGNPFQFYLT RVSGVKPKYN SGALHIKDIL SPLFGTLVSS AQFNYCFDVD WLVKQYPPEF RKKPILLVHG DKREAKAHLH AQAKPYENIS LCQAKLDIAF GTHHTKMMLL LYEEGLRVVI HTSNLIHADW HQKTQGTHL (1-298 aa encoded by BC006083 )
The protein encoded by this gene is involved in repairing stalled topoisomerase I-DNA complexes by catalyzing the hydrolysis of the phosphodiester bond between the tyrosine residue of topoisomerase I and the 3-prime phosphate of DNA. This protein may also remove glycolate from single-stranded DNA containing 3-prime phosphoglycolate, suggesting a role in repair of free-radical mediated DNA double-strand breaks. This gene is a member of the phospholipase D family and contains two PLD phosphodiesterase domains. Mutations in this gene are associated with the disease spinocerebellar ataxia with axonal neuropathy (SCAN1). While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same isoform.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tyr-DNA phosphodiesterase 1; Tyrosyl-DNA phosphodiesterase 1
Gene Aliases: TDP1
UniProt ID: (Human) Q9NUW8
Entrez Gene ID: (Human) 55775
Molecular Function: phosphodiesterase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support