Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 86%; Horse: 86%; Human: 100%; Pig: 86%
TDRD12 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ES cell associated transcript 8; ES cell-associated transcript 8 protein; Putative ATP-dependent RNA helicase TDRD12; Tudor domain-containing protein 12
Gene Aliases: ECAT8; TDRD12
UniProt ID: (Human) Q587J7
Entrez Gene ID: (Human) 91646
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support