Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSEQERIQE CLRKEIRSLL ISTKDGLSPQ ELEKEYLLMV GNHLPLRILG YRSTMELVLD MPDVVRVCPG AGGTVILKAI PDESTKGIAS LVAKQRSSHK LRNSMHKGRP SIYSGPRSHR RVPYRGSVAP ILPAVVKSEL KDLLALSPVL LSDFEKAFAK RFGRSFQYMQ YGFLSMFEVL NAASDVISVE QTRAGSLLML KKSVTEEKPR GCPAGKIFTQ PFRMKQGSYS TGFPVAKPCF SQPTSNMEPP KQIMSMEKTS KLNVVETSRL NHTEKLNQLE NTFKSVIAQI GPGGTISSEL KHKIKFVVSK FPEGLFISKL LGEYEVIFKE QLSPKKLGFL NVTELVGALS D (1-350 aa encoded by BC130532)
TDRD5, also named as Tudor domain-containing protein 5, is a 981 amino acid protein, which contains one Tudor domain and three HTH OST-type domains. TDRD5 localizes in the cytoplasm and belongs to the TDRD5 family. TDRD5 is required during spermiogenesis to participate in the repression transposable elements and prevent their mobilization, which is essential for the germline integrity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: testicular tissue protein Li 194; tudor domain containing protein 5; Tudor domain-containing protein 5
Gene Aliases: Gm103; TDRD5; TUDOR3
UniProt ID: (Human) Q8NAT2, (Mouse) Q5VCS6
Entrez Gene ID: (Human) 163589, (Mouse) 214575
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support