Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TDRKH is a protein of unknown function containing one tudor domain, and two KH domains. It has been found to interact with PIWIL1 and PIWIL4 by binding to methylated arginines residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: putative RNA binding protein; RP11-98D18.8; TDRD7; tudor and KH domain containing protein; Tudor and KH domain-containing protein; tudor domain containing 2; Tudor domain-containing protein 2
Gene Aliases: 2700091C21Rik; RGD1311196; TDRD2; TDRKH
UniProt ID: (Human) Q9Y2W6, (Mouse) Q80VL1
Entrez Gene ID: (Human) 11022, (Pig) 100622748, (Mouse) 72634, (Rat) 310652
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support