Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: DQVSKDKAF QTMATMSSAQ LISAPSLQAK LGPTGPQVVQ ASELFQFWSG GSGPPWNVPD VKPFSQTPFT LSLTPPSTDL PGYEPPQALS PLPPPTPSPP AWQARGLGTA RLQLVEFSAF VEPPDAVDSY QR (121-251 aa encoded by BC051301)
TEAD2 is a putative transcription factor that binds to the SPH and GT-IIC enhansons (5'-GTGGAATGT-3'). TEAD2 may be involved in the gene regulation of neural development. TEAD2 binds to the M-CAT motif.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Embryonic TEA domain-containing factor; ETEF-1; ETF; TEA domain family member 2; TEAD-2; Transcriptional enhancer factor TEF-4
Gene Aliases: Etdf; ETF; TEAD-2; TEAD2; TEF-4; TEF4
UniProt ID: (Human) Q15562, (Mouse) P48301
Entrez Gene ID: (Human) 8463, (Mouse) 21677
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support