Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SAQIISATA FHSSMALARG PGRPAVSGFW QGALPGQAGT SHDVKPFSQQ TYAVQPPLPL PGFESPAGPA PSPSAPPAPP WQGRSVASSK LWMLEFSAFL EQQQDPDTYN KHLFVHIGQS SPSYSDPYLE AVDIRQIYDK FPEKKGGLKD LFERGPSNAF FLVKFWADLN TNIEDEGSSF YGVSSQYESP ENMIITCSTK VCSFGKQVVE KVETEYARYE NGHYSYRIHR SPLCEYMINF IHKLKHLPEK YMMNSVLENF TILQVVTNRD TQETLLCIAY VFEVSASEHG AQHHIYRLVK E (135-434 aa encoded by BC015497)
This gene product is a member of the transcriptional enhancer factor (TEF) family of transcription factors, which contain the TEA/ATTS DNA-binding domain. It is preferentially expressed in the skeletal muscle, and binds to the M-CAT regulatory element found in promoters of muscle-specific genes to direct their gene expression. Alternatively spliced transcripts encoding distinct isoforms, some of which are translated through the use of a non-AUG (UUG) initiation codon, have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ETF-related factor 2; ETF-related factor-2; ETFR-2; FGF-regulated 19; related transcription enhancer factor 1B; RTEF-1; TEA domain family member 4; TEAD-4; TEF-1-related factor 1; TEF-1-related factor FR-19; Transcription factor 13-like 1; Transcription factor RTEF-1; transcriptional enhancer factor 1-related; transcriptional enhancer factor 3; Transcriptional enhancer factor TEF-3
Gene Aliases: EFTR-2; ETFR-2; Etfr2; FR-19; hRTEF-1B; RTEF1; TCF13L1; Tcf13r1; TEAD-4; TEAD4; TEF-3; TEF3; Tefr; TEFR-1; Tefr1; Tefr1a
UniProt ID: (Human) Q15561, (Mouse) Q62296
Entrez Gene ID: (Human) 7004, (Mouse) 21679, (Rat) 683522
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support