Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: KSTTPKKITP NVTFCDENAK EPENALDKLF SSEQQASILH VLNTASTKEL EAFRLLRGRR SINIVEHREN FGPFQNLESL MNVPLFKYKS TVQVCNSILC PKTGREKRKS PENRFLRKLL KPDIERERLK AVNSIISIVF GTRRIAWAHL DRKLTVLDWQ QSDRWSLMRG IYSSS; Positive Samples: HL-60, K-562, Mouse liver, Rat liver; Cellular Location: Mitochondrion matrix, mitochondrion nucleoid
TEFM is a transcription elongation factor which increases mitochondrial RNA polymerase processivity. It regulates transcription of the mitochondrial genome, including genes important for the oxidative phosphorylation machinery.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: transcription elongation factor of mitochondria; Transcription elongation factor, mitochondrial
Gene Aliases: 1110002N22Rik; C17orf42; RGD1306595; TEFM
UniProt ID: (Human) Q96QE5, (Mouse) Q5SSK3, (Rat) Q4KM51
Entrez Gene ID: (Human) 79736, (Mouse) 68550, (Rat) 287554
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support