Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MATLSVKPS RRFQLPDWHT NSYLLSTNAQ LQRDASHQIR QEARVLRNET NNQTIWDEHD NRTRLVERID TVNRWKEMLD KCLTDLDAEI DALTQMKESA EQNLQAKNLP LDVAIECLTL RESRRDIDVV KDPVEDELHK EVEVIEATKK ALQQKVSQAF EQLCLLQEVQ QQLNSDHRGK METLEIDRGC LSLNLRSPNI SLKVDPTRVP DGSTTLQQWD DFSRFNKDRA EAEMKAATEL REATALTIAE TNNELEAQRV ATEFAFRKRL REMEKVYSEL KWQEKNTLEE IAELQEDIRH LEEDLRTKLL SLKLSHTRLE ARTYRPNVEL CRDQAQYGLT DEVHQLEATI AALKQKLAQA QDALDALCKH LARLQADIAC KANSMLLDTK CMDTRRKLTV PAERFVPEVD TFTRTTNSTL SPLKSCQLEL A (1-430 aa encoded by BC035620)
Structural component of ciliary and flagellar microtubules. Plays a key role in the assembly or attachment of the inner dynein arm to microtubules in sperm flagella and tracheal cilia. Forms filamentous polymers in the walls of ciliary and flagellar microtubules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TEKTB1; tektin 2 (testicular); Tektin-2; tektin-B1; Tektin-t; Testicular tektin; Testicular tektin B1-like protein
Gene Aliases: AU014974; h-tektin-t; TEKT2; TEKTB1; TEKTIN-T
UniProt ID: (Human) Q9UIF3, (Mouse) Q922G7, (Rat) Q6AYM2
Entrez Gene ID: (Human) 27285, (Mouse) 24084, (Rat) 298532
Molecular Function: non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support