Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Structural component of ciliary and flagellar microtubules. Plays a key role in the assembly or attachment of the inner dynein arm to microtubules in sperm flagella and tracheal cilia. Forms filamentous polymers in the walls of ciliary and flagellar microtubules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TEKTB1; tektin 2 (testicular); Tektin-2; tektin-B1; Tektin-t; Testicular tektin; Testicular tektin B1-like protein
Gene Aliases: h-tektin-t; TEKT2; TEKTB1; TEKTIN-T
UniProt ID: (Human) Q9UIF3
Entrez Gene ID: (Human) 27285
Molecular Function: non-motor microtubule binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support