Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • TER-119 Antibodies

            Invitrogen

            TER-119 Monoclonal Antibody (TER-119), PE-Cyanine7, eBioscience™

            38 Published Figures
            86 References
            View all (39) TER-119 antibodies

            Promotions

            View available promotion(s)

            Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more

            Datasheet
            Protocols
            Questions & Answers
            Catalog # 25-5921-82
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            215.90
            Special offer
            Online exclusive
            254.00
            Save 38.10 (15%)
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite TER-119 Monoclonal Antibody (TER-119), PE-Cyanine7, eBioscience™

            • Antibody Testing Data (1)
            • Published Figures (38)
            Application
            TER-119 Antibody in Flow Cytometry (Flow)
            Group 53 Created with Sketch.

            FIGURE: 1 / 39

            TER-119 Antibody (25-5921-82) in Flow

            Staining of C57BL/6 bone marrow cells with Anti-Mouse CD45 FITC (Product # 11-0451-82) and 0.25 µg of Rat IgG2b K Isotype Control PE-Cyanine7 (Product # 25-4031-82) (left) or 0.25 µg of Anti-Mouse TER-119 PE-Cyanine7 (right). Total viable cells were used for analysis.
            View Product

            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)
            TER-119 Antibody in Flow Cytometry (Flow)

            Product Details

            25-5921-82

            Applications
            Tested Dilution
            Publications

            Flow Cytometry (Flow)

            0.5 µg/test
            View 86 publications 86 publications
            Product Specifications

            Species Reactivity

            Mouse

            Published species

            Fish, Human, Mouse

            Host/Isotype

            Rat / IgG2b, kappa

            Recommended Isotype Control

            Rat IgG2b kappa Isotype Control (eB149/10H5), PE-Cyanine7, eBioscience™

            Class

            Monoclonal

            Type

            Antibody

            Clone

            TER-119

            Conjugate

            PE-Cyanine7
            • Unconjugated
            • Alexa Fluor 660 
            • Alexa Fluor 700 
            • APC 
            • APC-eFluor 780
            • Biotin 
            • Brilliant UV 395
            • Brilliant UV 615
            • Brilliant UV 661
            • Brilliant UV 737
            • Brilliant UV 805
            • Brilliant Violet 421
            • Brilliant Violet 480
            • eFluor 450
            • eFluor 506
            • FITC
            • Functional Grade
            • PE 
            • PE-Cyanine5
            • PE-Cyanine5.5
            • PE-eFluor 610
            • PerCP-Cyanine5.5
            • PerCP-eFluor 710
            • Super Bright 436
            • Super Bright 600
            • Super Bright 645
            • Super Bright 702
            • Super Bright 780
            • Request custom conjugation

            Excitation/Emission Max

            569/780 nm View spectra spectra

            Form

            Liquid

            Concentration

            0.2 mg/mL

            Purification

            Affinity chromatography

            Storage buffer

            PBS, pH 7.2

            Contains

            0.09% sodium azide

            Storage conditions

            4°C, store in dark, DO NOT FREEZE!

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            RRID

            AB_469661

            Product Specific Information

            Description: The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin.

            Applications Reported: This TER-119 antibody has been reported for use in flow cytometric analysis.

            Applications Tested: This TER-119 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

            Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light.

            Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 µL cell sample + 100 µL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

            Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.

            Filtration: 0.2 µm post-manufacturing filtered.

            Target Information

            TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            How to build a flow cytometry experiment with Invitrogen Flow Cytometry Panel Builder
            The Invitrogen Flow Cytometry Panel Builder webtool helps you chose antibody clones, fluorophores, fluorescent proteins, and reagents to design your experiment.
            Video Player is loading.
            Current Time 0:00
            /
            Duration 0:00
            Loaded: 0%
            0:00
            Stream Type LIVE
            Remaining Time -0:00
             
            1x
            • 2x
            • 1.75x
            • 1.5x
            • 1.25x
            • 1x, selected
            • 0.75x
            • 0.5x
            • Chapters
            • descriptions off, selected
            • captions settings, opens captions settings dialog
            • captions off, selected

              This is a modal window.

              Beginning of dialog window. Escape will cancel and close the window.

              Text
              Text Background
              Caption Area Background
              Font Size
              Text Edge Style
              Font Family

              End of dialog window.

              This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.

              How to use the Panel Builder

              Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

              Bioinformatics

              Protein Aliases: Ly-76; Ly76; TER119

              View more View less

              It has to be done as per old AB suggested Products section.
              guarantee_icon

              Performance Guarantee

              If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

              Learn more
              help_icon

              We're here to help

              Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

              Contact tech support
              Ordering Plus Icon Minus Icon
              • Order Status
              • Order Help
              • Quick Order
              • Supply Center
              • eProcurement
              Support Plus Icon Minus Icon
              • Help and Support
              • Contact Us
              • Technical Support Centers
              • Documents and Certificates
              • Report a Site Issue
              Resources Plus Icon Minus Icon
              • Learning Centers
              • Promotions
              • Events and Webinars
              • Social Media
              About Thermo Fisher Plus Icon Minus Icon
              • About Us
              • Careers
              • Investors
              • News
              • Social Responsibility
              • Trademarks
              • Consumer Health Data Privacy Policy
              Our Portfolio Plus Icon Minus Icon
              • Thermo Scientific
              • Applied Biosystems
              • Invitrogen
              • Gibco
              • Ion Torrent
              • Fisher Scientific
              • Unity Lab Services
              • Patheon
              • PPD
              • Terms & Conditions
              • Privacy Information Center
              • Price & Freight Policy
              • Cookie Preferences

                Your choices regarding cookies on this site

                We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
              © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
              united states flag icon
              United States

              Your items have has been added!


              Host server : magellan-search-green-56756dfb8b-l9gpq:80/100.66.77.183:80.
              git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
              git-url: https://github.com/thermofisher/magellan-search
              git-branch: release/2.31.1-2025.06.25-1.0