Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: GPGTMPLADW QEPLAPPIRR WCSLPGSPEF LHQEACPFVG REESLSDGPP PRLSSLKYRV KEIPPFRASA LPAAQAHEAM DCSILQEENG FGSRPQGTSP CPAGASEEME VEERPAGSTP ATFSTSGIGL QTQGKQDG
TESK2 or testis-specific kinase 2 is a serine/threonine protein kinase that contains an N-terminal protein kinase domain and belongs to the TESK subgroup of the LIMK/TESK family of protein kinases. The kinase domain of TESK2 is structurally similar to the kinase domain of testis-specific protein kinase-1 and the LIM motif-containing protein kinases (LIMKs). TESK2 is predominantly expressed in testis and prostate and plays an important role in spermatogenesis. TESK2 phosphorylates cofilin specifically at Ser-3 and induces formation of actin stress fibers and focal adhesions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dual specificity testis-specific protein kinase 2; Testicular protein kinase 2
Gene Aliases: TESK2
UniProt ID: (Human) Q96S53
Entrez Gene ID: (Human) 10420
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support