Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MVLKAEHTRS PSATLPSNVP SCRSLSSSED GPSGPSSLAD GGLAHNLQDS VRHRILYLSE QLRVEKASRD GNTVSYLKLV SKADRHQVPH IQQAFEKVNQ RASATIAQIE HRLHQCHQQL QELEEGCRPE GLLLMAESDP ANCEPPSEKA LLSEPPEPGG EDGPVNLPHA SRPFILESRF QSLQQGTCLE TEDVAQQQNL LLQKVKAELE EAKRFHISLQ ESYHSLKERS LTDLQLLLES LQEEKCRQAL MEEQVNGRLQ GQLNEIYNLK HNLACSEERM AYLSYERAKE IWEITETFKS RISKLEMLQQ VTQLEAAEHL QSRPPQMLFK FLSPRLSLAT VLLVFVSTLC ACPSSLISSR LCTCTMLMLI GLGVLAWQRW RAIPATDWQE WVPSRCRLYS KDSGPPADGP
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Testis-specific protein TEX28
Gene Aliases: CXorf2; fTEX; MRX99; TEX28; TEX28P1; TEX28P2
UniProt ID: (Human) O15482
Entrez Gene ID: (Human) 1527
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support