Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: LHAQPQPQH PGWPGQRQSQ ESGLLHTHRG LPHQLSGLDP RRDYRRHEDL LHGPHALSSG LGDLSIHSLP HAIEEVPHVE DPGINIPDQT VIKKGPVSLS KSNSNAVSAI PINKDNLFGG VVNPNEVFCS VPGRLSLLSS TSKYKVTVAE VQRRLSPPEC LNASLLGGVL RRAKSKNGGR SLREKLDKIG LNLPAGRRKA ANVTLLTSLV EGEAVHLARD FGYVCETEFP AKAVAEFLNR QHSDPNEQVT RKNMLLATKQ ICKEFTDLLA QDRSPLGNSR PNPILEPGIQ SCLTHFNLIS HGFGSPAVCA AVTALQNYLT EALKAMDKMY LSNNPNSHTD NNAKSSDKEE KHRK (79-431 aa encoded by BC017754)
TFAP2A is a sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limbs and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC. TFAP2A is the only AP-2 protein required for early morphogenesis of the lens vesicle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating enhancer-binding protein 2-alpha; Activator protein 2; AP-2; AP-2 transcription factor; AP2-alpha; ERF-1; FLJ51761; OTTHUMP00000214240; OTTHUMP00000214243; transcription factor AP-2 alpha (activating enhancer binding protein 2 alpha); Transcription factor AP-2-alpha
Gene Aliases: AP-2; AP-2alpha; AP2TF; BOFS; TFAP2; TFAP2A
UniProt ID: (Human) P05549
Entrez Gene ID: (Human) 7020
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support