Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GRNEMAARKN MLLAAQQLCK EFTELLSQDR TPHGTSRLAP VLETNIQNCL SHFSLITHGF GSQAICAAVS ALQNYIKEAL IVIDKSYMNP GDQSPADSNK TLEKMEKHRK
The protein encoded by this gene is a sequence-specific DNA-binding transcription factor involved in the activation of several developmental genes. The encoded protein can act as either a homodimer or heterodimer with other family members and is induced during retinoic acid-mediated differentiation. It plays a role in the development of the eyes, face, body wall, limbs, and neural tube.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating enhancer-binding protein 2 gamma; AP2-gamma; estrogen receptor factor 1; Transcription factor AP-2 gamma; transcription factor AP-2 gamma (activating enhancer binding protein 2 gamma); transcription factor AP-2 gamma (activating enhancer-binding protein 2 gamma); Transcription factor ERF-1
Gene Aliases: AP2-GAMMA; ERF1; hAP-2g; TFAP2C; TFAP2G
UniProt ID: (Human) Q92754
Entrez Gene ID: (Human) 7022
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support