Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MAASGKLSTC RLPPLPTIRE IIKLLRLQAA KQLSQNFLLD LRLTDKIVRK AGNLTNAYVY EVGPGPGGIT RSILNADVAE LLVVEKDTRF IPGLQMLSDA APGKLRIVHG DVLTFKVEKA FSESLKRPWE DDPPNVHIIG NLPFSVSTPL IIKWLENISC RDGPFVYGRT QMTLTFQKEV AERLAANTGS KQRSRLSVMA QYLCNVRHIF TIPGQAFVPK PEVDVGVVHF TPLIQPKIEQ PFKLVEKVVQ NVFQFRRKYC HRGLRMLFPE AQRLESTGRL LELADIDPTL RPRQLSISHF KSLCDVYRKM CDEDPQLFAY NFREELKRRK SKNEEKEEDD AENYRL
The protein encoded by this gene is a dimethyltransferase that methylates the conserved stem loop of mitochondrial 12S rRNA. The encoded protein also is part of the basal mitochondrial transcription complex and is necessary for mitochondrial gene expression. The methylation and transcriptional activities of this protein are independent of one another. Variations in this gene may influence the severity of aminoglycoside-induced deafness.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Dimethyladenosine transferase 1, mitochondrial; h-mtTFB; h-mtTFB1; homolog of yeast mitochondrial transcription factor B; hTFB1M; Mitochondrial 12S rRNA dimethylase 1; mitochondrial dimethyladenosine transferase 1; Mitochondrial transcription factor B1; OTTHUMP00000017473; S-adenosylmethionine-6-N', N'-adenosyl(rRNA) dimethyltransferase 1
Gene Aliases: CGI-75; CGI75; mtTFB; mtTFB1; TFB1M
UniProt ID: (Human) Q8WVM0
Entrez Gene ID: (Human) 51106
Molecular Function: RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support