Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: YSMSDVLALP IFKQEESSLP PDNENKILPF QYVLCAATSP AVKLHDETLT
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 93%
This gene encodes a transcription factor that binds the alpha-globin promoter and activates transcription of the alpha-globin gene. The encoded protein regulates erythroid gene expression, plays a role in the transcriptional switch of globin gene promoters, and it activates many other cellular and viral gene promoters. The gene product interacts with certain inflammatory response factors, and polymorphisms of this gene may be involved in the pathogenesis of Alzheimer's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha-globin transcription factor CP2; Late SV40 factor; LBP 1C; SAA3 enhancer factor; Transcription factor LSF
Gene Aliases: LBP1C; LSF; LSF1D; SEF; TFCP2; TFCP2C
UniProt ID: (Human) Q12800
Entrez Gene ID: (Human) 7024
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support