Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLFWHTQPEH YNQHNSGSYL RDVLALPIFK QEEPQLSPEN EARLPPLQYV LCAATSPAVK LHEETLTYLN QGQSYEIRLL ENRKLGDFQD LNTKYVKSII RVVFHDRRLQ YTEHQQLEGW RWSRPGDRIL DIDIPLSVGI LDPRASPTQL NAVEFLWDPA KRASAFIQVH CISTEFTPRK HGGEKGVPFR VQIDTFKQNE NGEYTEHLHS ASCQIKVFKP KGADRKQKTD REKMEKRTAQ EKEKYQPSYE TTILTECSPW PDVAYQVNSA PSPSYNGSPN SFGLGEGNAS PTHPVEALPV GSDHLLPSAS IQDAQQWLHR NRFSQFCRLF ASFSGADLLK MSRDDLVQIC GPADGIRLFN AIKGRNVRPK MTIYVCQELE QNRVPLQQKR DGSGDSNLSV YHAIFLEELT TLELIEKIAN LYSISPQHIH RVYRQGPTGI HVVVSNEMVQ NFQDESCFVL STIKAESNDG YHIILKCGL
TFCP2L1 is a candidate CP2 family member. It is expressed in a developmentally regulated fashion in vivo and acts as a direct repressor of transcription. CP2-related proteins comprise a family of DNA-binding transcription factors that are generally activators of transcription and expressed ubiquitously.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CP2-related transcriptional repressor 1; CRTR-1; Transcription factor CP2-like protein 1; Transcription factor LBP-9
Gene Aliases: CRTR1; LBP-9; LBP9; TFCP2L1
UniProt ID: (Human) Q9NZI6
Entrez Gene ID: (Human) 29842
Molecular Function:
DNA-binding transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support