Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MSLSHLYRDG EGRIDDDDDE RENFEITDWD LQNEFNPNRQ RHWQTKEEAT YGVWAERDSD DERPSFGGKR ARDYSAPVNF ISAGLKKGAA EEAELEDSDD EEKPVKQDDF PKDFGPRKLK TGGNFKPSQK GFAGGTKSFM; Positive Samples: LO2, HeLa, A-431, MCF-7, HT-1080, Mouse brain, Mouse liver; Cellular Location: Cytoplasm, Nucleus
Involved in pre-mRNA splicing, specifically in spliceosome disassembly during late-stage splicing events. Intron turnover seems to proceed through reactions in two lariat-intron associated complexes termed Intron Large (IL) and Intron Small (IS). In cooperation with DHX15 seems to mediate the transition of the U2, U5 and U6 snRNP-containing IL complex to the snRNP-free IS complex leading to efficient debranching and turnover of excised introns. May play a role in the differentiation of ameloblasts and odontoblasts or in the forming of the enamel extracellular matrix.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Septin and tuftelin-interacting protein 1; STIP-1; Tuftelin-interacting protein 11; tuftelin-interacting protein 33; Tuftelin-interacting protein 39; tuftelin-interacting protein, 39 kD
Gene Aliases: 2810002G02Rik; AF097181; AW046167; bK445C9.6; hNtr1; HSPC006; NTR1; Spp382; Srr1; STIP; TFIP11; TIP33; TIP39
UniProt ID: (Human) Q9UBB9, (Mouse) Q9ERA6
Entrez Gene ID: (Human) 24144, (Mouse) 54723
Molecular Function:
RNA splicing factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support